Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174767.2 | NP_777192.1 | 3 - 4 | 666 | 57 | |
GenBank mRNA | AF251032.1 | - | 3 - 4 | 666 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |