Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFDP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174800.2 | NP_777225.1 | 5 - 6 | 1609 | 112 | |
GenBank mRNA | D84513.1 | - | 5 - 6 | 1609 | 112 | |
D84514.1 | - | 1142 | 112 | |||
D84515.1 | - | 127 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Bos taurus Probe spans exons |