Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATHL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174826.3 | NP_777251.1 | 3 - 4 | 390 | 197 | |
GenBank mRNA | BC120477.1 | - | 3 - 4 | 396 | 197 | |
L02650.1 | - | 3 - 4 | 393 | 197 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |