Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174835.2 | NP_777260.1 | 5 - 6 | 635 | 73 | |
GenBank mRNA | BC116120.1 | - | 5 - 6 | 638 | 73 | |
U21660.1 | - | 5 - 6 | 635 | 73 | ||
Z50026.1 | - | 645 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |