Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175782.1 | NP_786976.1 | 3 - 4 | 298 | 66 | |
GenBank mRNA | AB098790.1 | - | 1 - 2 | 209 | 66 | |
AB098791.1 | - | 1 - 2 | 210 | 66 | ||
AB098800.1 | - | 1 - 2 | 210 | 66 | ||
AB099023.1 | - | 3 - 4 | 308 | 66 | ||
AB099039.1 | - | 3 - 4 | 306 | 66 | ||
AB099046.1 | - | 3 - 4 | 305 | 66 | ||
BC103156.1 | - | 3 - 4 | 323 | 66 | ||
X14330.1 | - | 298 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |