Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176609.3 | NP_788782.2 | 1 - 2 | 94 | 63 | |
GenBank mRNA | BC122610.1 | - | 1 - 2 | 46 | 63 | |
DV824401.1 | - | 1 - 2 | 93 | 63 | ||
EH148795.1 | - | 1 - 2 | 126 | 63 | ||
X56685.1 | - | 43 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |