Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UMPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177508.1 | NP_803474.1 | 1 - 2 | 195 | 144 | |
GenBank mRNA | BC112872.1 | - | 1 - 2 | 193 | 144 | |
X65125.1 | - | 195 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |