Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181018.2 | NP_851361.1 | 2 - 3 | 216 | 81 | |
GenBank mRNA | AF001261.1 | - | 2 - 3 | 216 | 81 | |
AF001262.1 | - | 2 - 3 | 212 | 81 | ||
AF001263.1 | - | 2 - 3 | 347 | 81 | ||
AF001264.1 | - | 2 - 3 | 262 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Bos taurus Probe spans exons |