Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181022.2 | NP_851365.1 | 1 - 2 | 245 | 88 | |
GenBank mRNA | K03253.1 | - | 1 - 2 | 172 | 88 | |
K03254.1 | - | 1 - 2 | 201 | 88 | ||
X02440.1 | - | 245 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |