Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205792.2 | NP_991361.1 | 5 - 6 | 1299 | 142 | |
NM_205814.2 | NP_991383.1 | 5 - 6 | 1299 | 142 | ||
GenBank mRNA | AB114473.1 | - | 5 - 6 | 1299 | 142 | |
AB114474.1 | - | 5 - 6 | 1299 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |