Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205794.1 | NP_991363.1 | 8 - 9 | 1032 | 60 | |
GenBank mRNA | AY533678.1 | - | 8 - 9 | 1032 | 60 | |
BC151491.1 | - | 8 - 9 | 1140 | 60 | ||
BT030588.1 | - | 8 - 9 | 1093 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |