Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAPTM4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205802.2 | NP_991371.2 | 5 - 6 | 742 | 122 | |
GenBank mRNA | AF276819.1 | - | 5 - 6 | 693 | 122 | |
BC149605.1 | - | 5 - 6 | 740 | 122 | ||
EV638847.1 | - | 5 - 6 | 451 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |