Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205803.1 | NP_991372.1 | 3 - 4 | 308 | 74 | |
GenBank mRNA | AJ012589.1 | - | 3 - 4 | 308 | 74 | |
BC102657.1 | - | 3 - 4 | 321 | 74 | ||
BC151412.1 | - | 2 - 3 | 270 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |