Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001134.1 | NP_001001134.1 | 5 - 6 | 1184 | 69 | |
GenBank mRNA | AJ508719.1 | - | 5 - 6 | 1184 | 69 | |
BC123736.1 | - | 5 - 6 | 1183 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |