Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002884.1 | NP_001002884.1 | 3 - 4 | 323 | 68 | |
GenBank mRNA | AY156928.1 | - | 3 - 4 | 323 | 68 | |
BC149263.1 | - | 3 - 4 | 389 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |