Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSP90AA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012670.2 | NP_001012688.1 | 7 - 8 | 1484 | 109 | |
GenBank mRNA | AB072368.1 | - | 7 - 8 | 1451 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Bos taurus Probe spans exons |