Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013402.1 | NP_001013420.1 | 3 - 4 | 346 | 60 | |
GenBank mRNA | BC123798.1 | - | 3 - 4 | 343 | 60 | |
BT020746.1 | - | 3 - 4 | 326 | 60 | ||
BT020791.1 | - | 3 - 4 | 320 | 60 | ||
BT020797.1 | - | 3 - 4 | 346 | 60 | ||
BT020850.1 | - | 3 - 4 | 335 | 60 | ||
BT021165.1 | - | 3 - 4 | 344 | 60 | ||
BT021167.1 | - | 3 - 4 | 335 | 60 | ||
BT021594.1 | - | 3 - 4 | 432 | 60 | ||
BT026293.1 | - | 3 - 4 | 297 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |