Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAGLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013599.1 | NP_001013617.1 | 3 - 4 | 418 | 82 | |
GenBank mRNA | BT020965.1 | - | 3 - 4 | 418 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |