Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014894.1 | NP_001014894.1 | 1 - 2 | 47 | 110 | |
GenBank mRNA | BT021487.1 | - | 1 - 2 | 47 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Bos taurus Probe spans exons |