Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEIL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014917.2 | NP_001014917.2 | 6 - 7 | 897 | 132 | |
GenBank mRNA | BT021698.1 | - | 6 - 7 | 883 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |