Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014936.1 | NP_001014936.1 | 2 - 3 | 180 | 70 | |
GenBank mRNA | AY911340.1 | - | 1 - 2 | 176 | 70 | |
BC102548.1 | - | 1 - 2 | 101 | 70 | ||
BC147944.1 | - | 1 - 2 | 216 | 70 | ||
BT021037.1 | - | 2 - 3 | 180 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |