Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015572.1 | NP_001015572.1 | 4 - 5 | 361 | 90 | |
GenBank mRNA | BC102707.1 | - | 4 - 5 | 382 | 90 | |
BT021074.1 | - | 4 - 5 | 361 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |