Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASGEF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015601.1 | NP_001015601.1 | 8 - 9 | 968 | 76 | |
GenBank mRNA | BT020843.1 | - | 8 - 9 | 968 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |