Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015607.1 | NP_001015607.1 | 3 - 4 | 702 | 94 | |
GenBank mRNA | BC114866.1 | - | 3 - 4 | 721 | 94 | |
BT020956.1 | - | 3 - 4 | 702 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |