Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015667.3 | NP_001015667.1 | 5 - 6 | 1016 | 111 | |
GenBank mRNA | BC103444.1 | - | 5 - 6 | 1017 | 111 | |
BT020782.1 | - | 5 - 6 | 955 | 111 | ||
BT020954.1 | - | 5 - 6 | 954 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |