Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015671.1 | NP_001015671.1 | 1 - 2 | 260 | 63 | |
GenBank mRNA | BT020945.1 | - | 1 - 2 | 260 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |