Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017951.1 | NP_001017951.1 | 2 - 3 | 686 | 72 | |
GenBank mRNA | BC114171.1 | - | 2 - 3 | 691 | 72 | |
BT021782.1 | - | 2 - 3 | 686 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |