Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024499.1 | NP_001019670.1 | 6 - 7 | 745 | 76 | |
GenBank mRNA | BC118212.1 | - | 6 - 7 | 739 | 76 | |
BT020772.1 | - | 6 - 7 | 830 | 76 | ||
BT021699.1 | - | 6 - 7 | 745 | 76 | ||
BT030475.1 | - | 3 - 4 | 687 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |