Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMFG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024537.2 | NP_001019708.1 | 3 - 4 | 203 | 73 | |
GenBank mRNA | AY911328.1 | - | 3 - 4 | 204 | 73 | |
BC103101.1 | - | 3 - 4 | 195 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |