Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024562.1 | NP_001019733.1 | 4 - 5 | 676 | 63 | |
GenBank mRNA | BC126787.1 | - | 4 - 5 | 460 | 63 | |
BT021531.1 | - | 4 - 5 | 676 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |