Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024692.1 | NP_001019863.1 | 9 - 10 | 1432 | 151 | |
GenBank mRNA | AY965877.1 | - | 9 - 10 | 1432 | 151 | |
BC142234.1 | - | 9 - 10 | 1312 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |