Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TWF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025320.2 | NP_001020491.1 | 7 - 8 | 798 | 143 | |
GenBank mRNA | AY911376.1 | - | 7 - 8 | 799 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Bos taurus Probe spans exons |