Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031763.1 | NP_001026933.1 | 2 - 3 | 537 | 80 | |
GenBank mRNA | BT021524.1 | - | 2 - 3 | 537 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |