Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034249.2 | NP_001029421.1 | 5 - 6 | 630 | 61 | |
GenBank mRNA | BC102926.1 | - | 5 - 6 | 631 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |