Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034319.2 | NP_001029491.1 | 2 - 3 | 360 | 74 | |
GenBank mRNA | BC102927.1 | - | 2 - 3 | 361 | 74 | |
EF576938.1 | - | 2 - 3 | 345 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |