Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR2C2AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034327.1 | NP_001029499.1 | 3 - 4 | 604 | 117 | |
GenBank mRNA | BT021499.1 | - | 3 - 4 | 604 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Bos taurus Probe spans exons |