Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034381.2 | NP_001029553.1 | 3 - 4 | 135 | 87 | |
GenBank mRNA | BC102102.1 | - | 3 - 4 | 137 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |