Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034585.2 | NP_001029757.1 | 4 - 5 | 689 | 77 | |
GenBank mRNA | BC102992.1 | - | 4 - 5 | 690 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |