Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TECR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034748.2 | NP_001029920.1 | 10 - 11 | 747 | 107 | |
GenBank mRNA | BC102503.1 | - | 10 - 11 | 748 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |