Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX39A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034752.1 | NP_001029924.1 | 7 - 8 | 965 | 60 | |
GenBank mRNA | BC104616.1 | - | 8 - 9 | 979 | 60 | |
BT021050.1 | - | 7 - 8 | 965 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |