Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035070.2 | NP_001030242.2 | 8 - 9 | 1466 | 71 | |
GenBank mRNA | BC148874.1 | - | 8 - 9 | 1467 | 71 | |
BT021867.1 | - | 3 - 4 | 895 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |