Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035375.1 | NP_001030452.1 | 3 - 4 | 587 | 147 | |
GenBank mRNA | BC103026.1 | - | 3 - 4 | 587 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |