Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046591.2 | NP_001040056.2 | 3 - 4 | 646 | 133 | |
GenBank mRNA | BT020834.1 | - | 3 - 4 | 646 | 133 |
Gene Symbol: LOC107131181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC112860.1 | - | 4 - 5 | 589 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |