Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076498.2 | NP_001069966.1 | 5 - 6 | 458 | 78 | |
GenBank mRNA | BC102751.1 | - | 5 - 6 | 459 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |