Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CABP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174256.3 | NP_776681.1 | 3 - 4 | 602 | 86 | |
GenBank mRNA | AY048883.1 | - | 3 - 4 | 603 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |