Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174268.1 | NP_776693.1 | 2 - 3 | 185 | 148 | |
GenBank mRNA | AB081004.1 | - | 2 - 3 | 185 | 148 | |
AB846663.1 | - | 2 - 3 | 185 | 148 | ||
BC112462.1 | - | 2 - 3 | 249 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |