Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174471.3 | NP_776896.1 | 3 - 4 | 279 | 73 | |
GenBank mRNA | AF144763.1 | - | 2 - 3 | 127 | 73 | |
AY295346.1 | - | 3 - 4 | 279 | 73 | ||
BC102489.1 | - | 3 - 4 | 245 | 73 | ||
KX834564.1 | - | 2 - 3 | 197 | 73 | ||
M60073.1 | - | 2 - 3 | 203 | 73 | ||
S70841.1 | - | 2 - 3 | 199 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |