Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174486.3 | NP_776911.2 | 1 - 2 | 99 | 112 | |
GenBank mRNA | BC102904.1 | - | 1 - 2 | 145 | 112 | |
BT021018.1 | - | 1 - 2 | 99 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |