Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174535.2 | NP_776960.1 | 3 - 4 | 509 | 82 | |
GenBank mRNA | AB060107.1 | - | 1 | 384 | 82 | |
AB098991.1 | - | 1 | 369 | 82 | ||
AJ238405.1 | - | 2 - 3 | 321 | 82 | ||
BC105315.1 | - | 1 | 387 | 82 | ||
BC142302.1 | - | 1 | 389 | 82 | ||
DV888478.1 | - | 3 - 4 | 509 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |