Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174560.2 | NP_776985.2 | 3 - 4 | 515 | 78 | |
GenBank mRNA | AY072914.1 | - | 3 - 4 | 388 | 78 | |
BC149389.1 | - | 3 - 4 | 478 | 78 | ||
BT021838.1 | - | 3 - 4 | 515 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |